Order Entry
Belgium
ContactUsLinkComponent
PCB congener mix, method 8082A (19 components)
PCB congener mix, method 8082A (19 components)
Catalog # REST32416
Supplier:  Restek
CAS Number:  
undefined
PCB congener mix, method 8082A (19 components)
Catalog # REST32416
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    100 µg/ml each in isooctane
  • Pk:
    1 mL
  • Description:
    PCB congener mix, method 8082A
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Contains 19 components, those are 2-chlorobiphenyl , 2,3-dichlorobiphenyl , 2,2',5-trichlorobiphenyl , 2,4',5-trichlorobiphenyl, 2,2',3,5'-tetrachlorobiphenyl, 2,2',5,5'-tetrachlorobiphenyl, 2,3',4,4'-tetrachlorobiphenyl, 2,2',3,4,5'-pentachlorobiphenyl, 2,2',4,5,5'-pentachlorobiphenyl, 2,3,3',4',6-pentachlorobiphenyl, 2,2',3,4,4',5'-hexachlorobiphenyl, 2,2',3,4,5,5'-hexachlorobiphenyl, 2,2',3,5,5',6-hexachlorobiphenyl, 2,2',4,4',5,5'-hexachlorobiphenyl, 2,2',3,3',4,4',5-heptachlorobiphenyl, 2,2',3,4,4',5,5'-heptachlorobiphenyl, 2,2',3,4,4',5',6-heptachlorobiphenyl, 2,2',3,4',5,5',6-heptachlorobiphenyl and 2,2',3,3',4,4',5,5',6-nonachlorobiphenyl.

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.