Order Entry
Belgium
ContactUsLinkComponent
FAME #20 mix (8 components)
FAME #20 mix (8 components)
Catalog # REST35026
Supplier:  Restek
CAS Number:  
undefined
FAME #20 mix (8 components)
Catalog # REST35026
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    Neat
  • Pk:
    1 mL
  • Description:
    FAME #20 mix
  • Pk Info:
    50 mg/ampoule

Specifications

About this item

This mixture can be used for quantification (AOCS Method CE 1a–13) and approximate the compositions of the types of oils like coconut, palm kernel, babassu, and ouri-curi oil.

This mix contains eight components such as C8:00 methyl caprylate (7%), C10:00 methyl caprate (5%), C12:00 methyl laurate (48%), C14:00 methyl myristate (15%), C16:00 methyl palmitate (7%), C18:00 methyl stearate (3%), C18:01 methyl oleate (12%), C18:02 methyl linoleate (3%).

% represents weight/weight% basis (minimum 50 mg/ampoule).