Organochlorine pesticide mix AB #1 (20 components)
Catalog # REST32291
Supplier: Restek
CAS Number:
Specifications
- Concentration:200 µg/ml each in hexane:toluene (1:1)
- Pk:1 mL
- Description:Organochlorine pesticide mix AB #1
- Pk Info:1 ml/ampoule
Specifications
About this item
This mix contains 20 components.
Components are aldrin, α-BHC, β-BHC, δ-BHC, γ-BHC, cis-chlordane, trans-chlordane, 4,4'-DDD, 4,4'-DDE, 4,4'-DDT, dieldrin, endosulphan I, endosulphan II, endosulphan sulphate, endrin, endrin aldehyde, endrin ketone, heptachlor, heptachlor epoxide (isomer B), methoxychlor.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.