Order Entry
Belgium
ContactUsLinkComponent
Fuel oil degradation mix (4 components)
Fuel oil degradation mix (4 components)
Catalog # REST31240
Supplier:  Restek
CAS Number:  
undefined
Fuel oil degradation mix (4 components)
Catalog # REST31240
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 μg/ml each in methylene chloride
  • Pk:
    1 mL
  • Description:
    Fuel oil degradation mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Subsurface degradation of fuel oil spills can be estimated by examining the ratios of C17/pristane and C18/phytane. To assist in identifying these four compounds from the complex fuel oil analysis, Restek offer a product that contains these compounds for retention time determination.

Fuel oil degradation mix contains (C17) heptadecane, (C18) octadecane, pristane (2,6,10,14-tetramethylpentadecane), phytane (2,6,10,14-tetramethylhexadecane).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.