Order Entry
Belgium
ContactUsLinkComponent
8270 calibration mix #6 (10 components)
8270 calibration mix #6 (10 components)
  REST31623
 :  Restek
 :  
undefined
8270 calibration mix #6 (10 components)
  REST31623
 :  Restek
 :  

 

  • Concentration:
    2000 µg/ml each in methylene chloride
  • Pk:
    1 mL
  • Description:
    8270 calibration mix #6
  • Pk Info:
    1 ml/ampoule

 

 

Contains: Diallate (cis & trans), dimethoate, disulfoton, famphur, methyl parathion, parathion (ethyl parathion), phorate, propyzamide, O,O,O-triethyl phosphorothioate, zinophos (thionazine).

 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.