Method 525.2 surrogate standard mix
Catalog # REST31826
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 µg/ml each in acetone
- Pk:1 Ampoul
- Description:Method 525.2 surrogate standard mix
- Environmentally Preferable:
- Pk Info:1 ml/ampoule
Specifications
About this item
This mix contains 2-nitro-m-xylene, perylene-d12, pyrene-d10 and triphenylphosphate.