Order Entry
Belgium
Orders LinkContactUsLinkComponent
Lab control sample #2, L/C VOA

Lab control sample #2, L/C VOA

Lab control sample #2, 2500 μg/ml in P and T methanol, 1 ml/ampoule, 1 mL
Catalog # REST30093
Supplier:   Restek
Lab control sample #2, L/C VOA
Catalog # REST30093
Supplier:  Restek
CAS Number:  
Lab control sample #2, L/C VOA
Lab control sample #2, L/C VOA

Lab control sample #2, 2500 μg/ml in P and T methanol, 1 ml/ampoule, 1 mL

CAS Number:  
Price
56,20 €
Log in for availability
1 * 1 mL
Product Details & Documents

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.

Specifications

Specifications

  • Concentration:
    2500 μg/ml in P and T methanol
  • Pk:
    1 mL
  • Description:
    Lab control sample #2
  • Pk Info:
    1 ml/ampoule
  • Cat. No.:
    REST30093
  • Formula:
    C₂H₃Cl
  • CAS No.:
    75-01-4

Specifications