DRO mix (Tennessee/Mississippi) (16 components)
REST31214
: Restek
:
- Pk:1
- Description:DRO mix (Tennessee/Mississippi), 1000 μg/ml each in methylene chloride
- Pk Info:1 ml/ampoule
DRO mix contains (C10) n-decane, (C11) n-undecane, (C12) n-dodecane, (C13) n-tridecane, (C14) n-tetradecane, (C15) n-pentadecane, (C16) n-hexadecane, (C17) n-heptadecane, (C18) n-octadecane, (C19) n-nonadecane, (C20) n-eicosane, (C21) n-heneicosane, (C23) n-tricosane, (C24) n-tetracosane, (C25) n-pentacosane.
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.