Carbohydrate LC performance check mix (5 components)
Catalog # REST31809
Supplier: Restek
CAS Number:
Specifications
- Pk:1 Ampoul
- Description:Carbohydrate LC performance check mix, reconstitute in 1 ml acetonitrile:water (75:25) to 2,1, 2,0, 4,4, 4,5, 4,0 mg/ml, respectively
- Pk Info:Dry components in 4 ml screw-cap vial
Specifications
About this item
This carbohydrate LC performance check mix consist of five components which performance qualification (PQ) determines the precision of the LC system.
- For simple sugar analysis by LC with refractive index/light scattering detection
- Use with a silica column with amino functionality, as recommended by AOAC International
- Use as a calibration standard for AACC Method 80-04
- Offered dry for enhanced stability
Check mix for LC-RI consists of five simple sugars in varied concentrations: Glucose (2,1 mg), Fructose (2,0 mg), Lactose (4,4 mg), Maltose (4,5 mg), Sucrose (4,0 mg).
Performance qualification (PQ) determines the precision of the LC system. Our performance check mix for LC-RI consists of five simple sugars in varied concentrations. We prepare the reference material in water, dehydrate it, and package it dry for enhanced stability.