VOA calibration mix #1 (ketones) (4 components)
Catalog # REST30006
Supplier: Restek
CAS Number:
Specifications
- Concentration:5000 μg/ml each in P and T methanol:water (90:10)
- Pk:1
- Description:VOA calibration mix #1 (ketones)
- Pk Info:1 ml/ampoule
Specifications
About this item
Mix contains Acetone (67-64-1), 2-Butanone (MEK) (78-93-3), 2-Hexanone (591-78-6), 4-Methyl-2-pentanone (MIBK) (108-10-1).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.