C50 in toluene
Catalog # REST31685
Supplier: Restek
CAS Number:
Specifications
- Concentration:10 µg/ml in toluene
- Pk:1
- CAS No.:6596-40-3
- Description:n-Pentacontane (C50)
- Formula:C₅₀H₁₀₂
- Pk Info:1 ml/ampoule
Specifications
About this item
It has a shelf life 6 to 84 months.
- Meets CCME 2001 petroleum hydrocarbons in soil method-Tier 1
- Primary reference calibration standards for quantification of four fractions
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.