ContactUsLinkComponent
Chlorinated acids by LC, mix A (8 components)
  REST32431
 :  Restek
 :  
undefined
Chlorinated acids by LC, mix A (8 components)
  REST32431
 :  Restek
 :  
  • Pk:
    1 mL
  • Description:
    Chlorinated acids by LC, mix A, 1000 μg/ml each in acetonitrile
  • Pk Info:
    1 ml/ampoule
Consists of eight components, those are acifluorfen, bentazon, chloramben, 2,4-d, dicamba, dichlorprop, picloram and 2,4,5-tp.
 : Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.