CLP 04.1 B/N matrix spike mix (4 components)
Catalog # REST31492
Supplier: Restek
CAS Number:
Specifications
- Pk:1
- Description:CLP 04.1 B/N matrix spike mix, 1000 μg/ml each in methanol
- Pk Info:1 ml/ampoule
Specifications
About this item
The mix consists of acenaphthene, 2,4-dinitrotoluene, N-nitroso-di-n-propylamine, pyrene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.