ContactUsLinkComponent
8095 calibration mix A (10 components)
8095 calibration mix A (10 components)
Catalog # REST31607
Supplier:  Restek
CAS Number:  
undefined
8095 calibration mix A (10 components)
Catalog # REST31607
Supplier:  Restek
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    8095 calibration mix A, 1000 μg/ml each in acetonitrile
  • Pk Info:
    1 ml/ampoule
Specifications
About this item
The mix consists of, 2-amino-4,6-dinitrotoluene, 4-amino-2,6-dinitrotoluene, 1,3-dinitrobenzene, 2,4-dinitrotoluene, 2,6-dinitrotoluene, HMX, RDX, tetryl, 1,3,5-trinitrobenzene, 2,4,6-trinitrotoluene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.