8095 Matrix spike mix B (7 components)
Catalog # REST31610
Supplier: Restek
CAS Number:
Specifications
- Pk:1 KIT
- Description:8095 matrix spike mix B, 1000 μg/ml each in acetonitrile (3,5-dinitroaniline at 200 μg/ml)
- Pk Info:1 ml/ampoule
Specifications
About this item
The mix consists of, 3,5-dinitroaniline, nitrobenzene, nitroglycerin, 2-nitrotoluene, 3-nitrotoluene, 4-nitrotoluene, PETN.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.