Order Entry
Belgium
ContactUsLinkComponent
Pesticide standard mix B (11 components)
Pesticide standard mix B (11 components)
Catalog # REST32298
Supplier:  Restek
CAS Number:  
undefined
Pesticide standard mix B (11 components)
Catalog # REST32298
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    In hexane:toluene (90:10)
  • Pk:
    1
  • Description:
    Pesticide standard mix B
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Consists of 11 components, those are aldrin, β-bhc, δ-bhc, cis-chlordane, trans-chlordane, 4,4'-dde, endosulfan ii, endosulfan sulfate, endrin aldehyde, endrin ketone and heptachlor epoxide.

Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.