MA fractionation surrogate spike mix (2 components)
REST31480
: Restek
:
- Concentration:4000 µg/ml each in hexane
- Pk:1 mL
- Description:MA fractionation surrogate spike mix
- Pk Info:1 ml/ampoule
This MA fractionation surrogate spike mix contains two components: 2-Bromonaphthalene and 2-Fluorobiphenyl.
Shelf life 6 months.
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.