Order Entry
Belgium
ContactUsLinkComponent
European pharmacopoeia/ICH Class 2 mix B, revised (10 components)
European pharmacopoeia/ICH Class 2 mix B, revised (10 components)
Catalog # REST36270
Supplier:  Restek
CAS Number:  
undefined
European pharmacopoeia/ICH Class 2 mix B, revised (10 components)
Catalog # REST36270
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    Prepared in water:dimethyl sulphoxide (80:20)
  • Pk:
    1
  • Description:
    European pharmacopoeia/ICH Class 2 mixture B, revised
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The european pharmacopoeia/ICH Class 2 mix B, revised consists of 10 components, they are acetonitrile (410 μg/ml), chloroform (60 μg/ml), 1,2-dimethoxyethane (100 μg/ml), N,N-dimethylacetamide (1,090 μg/ml), 1,4-dioxane (380 μg/ml), 2-hexanone (50 μg/ml), methanol (3,000 μg/ml), nitromethane (50 μg/ml), pyridine (200 μg/ml) and tetralin (100 μg/ml).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.