ContactUsLinkComponent
EP 2.4.22 composition of fatty acids by GC mix 1 (6 components)
EP 2.4.22 composition of fatty acids by GC mix 1 (6 components)
Catalog # REST35100
Supplier:  Restek
CAS Number:  
undefined
EP 2.4.22 composition of fatty acids by GC mix 1 (6 components)
Catalog # REST35100
Supplier:  Restek
CAS Number:  
Specifications
  • Pk:
    1
  • Description:
    EP 2.4.22 composition of fatty acids by GC mixture 1
  • Pk Info:
    100 mg total
Specifications
About this item
The composition of fatty acids by GC mix 1 consists of 6 components, they are (C20:0) methyl arachidate 40%, (C12:0) methyl dodecanoate 5%, (C14:0) methyl myristate 5%, (C18:1 [cis9]) methyl oleate 20%, (C16:0) methyl palmitate 10% and (C18:0) methyl stearate 20%.