Order Entry
Belgium
ContactUsLinkComponent
Residual solvents - Class 1 (5 components)
Residual solvents - Class 1 (5 components)
Catalog # REST36279
Supplier:  Restek
CAS Number:  
undefined
Residual solvents - Class 1 (5 components)
Catalog # REST36279
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    In dimethyl sulphoxide
  • Pk:
    1 Ampoul
  • Description:
    Residual solvents - Class 1
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The solvents consists of five components, they are benzene (10 mg/ml), carbon tetrachloride (20 mg/ml), 1,2-dichloroethane 25 mg/ml, 1,1-dichloroethene (40 mg/ml), and 1,1,1-trichloroethane (50 mg/ml).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.