ContactUsLinkComponent
European pharmacopoeia/ICH Class 1 mix , revised (5 components)
  REST36261
 :  Restek
 :  
undefined
European pharmacopoeia/ICH Class 1 mix , revised (5 components)
  REST36261
 :  Restek
 :  
  • Concentration:
    In water:dimethyl sulfoxide (80:20)
  • Pk:
    1 Ampoul
  • Description:
    European pharmacopoeia/ICH Class 1 mixture (revised)
  • Pk Info:
    1 ml/ampoule
The european pharmacopoeia/ICH Class 1 mix consists of five components, they are benzene (2 μg/ml), carbon tetrachloride (4 μg/ml), 1,2-dichloroethane (5 μg/ml), 1,1-dichloroethene (8 μg/ml) and 1,1,1-trichloroethane (10 μg/ml).
 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.