Order Entry
Belgium
ContactUsLinkComponent
CLP 04.1 VOA internal standard/SMC spike mix (6 components)
CLP 04.1 VOA internal standard/SMC spike mix (6 components)
Catalog # REST30457
Supplier:  Restek
CAS Number:  
undefined
CLP 04.1 VOA internal standard/SMC spike mix (6 components)
Catalog # REST30457
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2500 µg/ml each in P and T methanol
  • Pk:
    1 mL
  • Description:
    CLP 04.1 VOA internal standard/SMC spike mixture
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

The VOA internal standard/SMC spike mix consists of 6 components, they are bromochloromethane,1-bromo-4-fluorobenzene (BFB), chlorobenzene-d5, 1,2-dichloroethane-d4, 1,4-difluorobenzene and toluene-d8.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.