European pharmacopoeia/ICH Class 1 mix (5 components)
Catalog # REST36228
Supplier: Restek
CAS Number:
Specifications
- Concentration:Prepared in water:dimethyl sulphoxide (80:20)
- Pk:1
- Description:European pharmacopoeia/ICH Class 1 mixture
- Pk Info:1 ml/ampoule
Specifications
About this item
The european pharmacopoeia/ICH Class 1 mix consists of five components, they are benzene (2 μg/ml), carbon tetrachloride (4 μg/ml), 1,2-dichloroethane (5 μg/ml), 1,1-dichloroethene (8 μg/ml) and 1,1,1-trichloroethane (1,500 μg/ml).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.