Order Entry
Belgium
ContactUsLinkComponent
CARB 1004 aldehyde/ketone-DNPH calibration standard (13 components)
CARB 1004 aldehyde/ketone-DNPH calibration standard (13 components)
Catalog # REST33093
Supplier:  Restek
CAS Number:  
undefined
CARB 1004 aldehyde/ketone-DNPH calibration standard (13 components)
Catalog # REST33093
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    3 µg/ml each in acetonitrile
  • Pk:
    1 mL
  • Description:
    Carb 1004 aldehyde/ketone DNPH calibration standard
  • Pk Info:
    1 ml/ampoule*

Specifications

About this item

The CARB 1004 aldehyde/ketone-DNPH calibration standard consists of 13 components, they are acetaldehyde-2,4-DNPH, acetone-2,4-DNPH, acrolein-2,4-DNPH, benzaldehyde-2,4-DNPH, n-butyraldehyde-2,4-DNPH, crotonaldehyde-2,4-DNPH, formaldehyde-2,4-DNPH, hexaldehyde-2,4-DNPH, methacrolein-2,4-DNPH, methyl ethyl ketone-2,4-DNPH, propionaldehyde-2,4-DNPH, m-tolualdehyde-2,4-DNPH, valeraldehyde-2,4-DNPH.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.