Tripentyltin chloride surrogate
Catalog # REST31477
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 μg/ml in methylene chloride
- Pk:1 mL
- Description:Tripentyltin chloride
- Pk Info:1 ml/ampul
Specifications
About this item
Complete data pack available for audit compliance.
This surrogate has a maximum shelf life of 36 months and a minimum of 6 months.