Tri-n-propyltin chloride surrogate
Catalog # REST31476
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml in methylene chloride
- Pk:1 mL
- Description:Tri-n-propyltin chloride
- Pk Info:1 ml/ampule
Specifications
About this item
Complete data pack available for audit compliance.