ContactUsLinkComponent
610 PAH calibration mix B (16 components)
  REST31455
 :  Restek
 :  
undefined
610 PAH calibration mix B (16 components)
  REST31455
 :  Restek
 :  
  • Concentration:
    In methylene chloride:methanol (1:1)
  • Pk:
    1 mL
  • Description:
    610 PAH calibration mix B
This 610 PAH calibration mix B idela for LC-UV detection.
It consists of 16 components: Acenaphthene (1000 μg/ml), Acenaphthylene (2000 μg/ml), Anthracene(100 μg/ml), Benz(a)anthracene (100 μg/ml), Benzo(a)pyrene (100 μg/ml), Benzo(b)fluoranthene (200 μg/ml), Benzo(k)fluoranthene (100 μg/ml), Benzo(ghi)perylene (200 μg/ml), Chrysene (100 μg/ml), Dibenz(a,h)anthracene (200 μg/ml), Fluoranthene (200 μg/ml), Fluorene (200 μg/ml), Indeno(1,2,3-cd)pyrene (100 μg/ml), Naphthalene (1000 μg/ml), Phenanthrene (100 μg/ml) and Pyrene (100 μg/ml).
 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.