ContactUsLinkComponent
GC multiresidue pesticide standard #2 (40 components)
GC multiresidue pesticide standard #2 (40 components)
Catalog # REST32564
Supplier:  Restek
CAS Number:  
undefined
GC multiresidue pesticide standard #2 (40 components)
Catalog # REST32564
Supplier:  Restek
CAS Number:  
Specifications
  • Pk:
    1 mL
  • Description:
    GC multiresidue pesticide standard #2, 100 μg/ml each in toluene
  • Pk Info:
    1 ml/ampoule
Specifications
About this item
Accurately identify and quantify pesticide residues by GC-MS/MS in fruits, vegetables, botanicals, and herbals such as tea, ginseng, ginger, echinacea and dietary supplements.
  • Quantitatively tested to confirm composition, detailed support documentation provided
  • Organochlorine compounds
Formulated and grouped for maximum long-term stability* and well-balanced chromatographic performance.
Optimised multiresidue pesticide method is offered free of charge and downloadable XLS file includes conditions and transition tables.
Consists of 40 components, those are aldrin, alpha-bhc, beta-bhc, delta-bhc, gamma-bhc , chlorbenside, cis-chlordane, trans-chlordane, chlorfenson , chloroneb, 2,4'-ddd, 4,4'-ddd, 2,4'-dde, 4,4'-dde, 2,4'-ddt, 4,4'-ddt, 4,4'-dichlorobenzophenone, dieldrin, endosulfan i, endosulfan ii, endosulfan ether, endosulfan sulfate, endrin, endrin aldehyde, endrin ketone, ethylan , fenson, heptachlor, heptachlor epoxide, hexachlorobenzene, isodrin, 2,4'-methoxychlor, 4,4'-methoxychlor olefin, mirex, cis-nonachlor, trans-nonachlor, pentachloroanisole, pentachlorobenzene, pentachlorothioanisole and tetradifon.
Certifications: Certified reference material (CRM) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.
Ordering information: The Comprehensive 203-compound kit also available (cat. # 32562) that covers food safety lists by the FDA, USDA, and other global governmental agencies; other ampoules also sold separately.
* NOTE: When combining many compounds with different chemical functionalities, mix stability can be an issue. In formulating these standards, we extensively studied the 203 compounds involved, and then grouped them into as few mixes as possible while still ensuring maximum long-term stability and reliability. For quantitative analysis, we recommend analysing each mix separately to ensure accurate results for every compound.