PCB kit #2
Catalog # REST32090
Supplier: Restek
CAS Number:
Specifications
- Concentration:200 µg/ml each in isooctane
- Pk:1
- Description:PCB kit #2
- Pk Info:1 ml/ampoule
Specifications
About this item
This kit contains Aroclor 1016, Aroclor 1221, Aroclor 1232, Aroclor 1242, Aroclor 1248, Aroclor 1254, Aroclor 1260.
- Aroclor standards may exhibit lot-to-lot variation
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.