Order Entry
Belgium
ContactUsLinkComponent
L/C aniline mix A (3 components)
L/C aniline mix A (3 components)
Catalog # REST31210
Fournisseur:  Restek
Numéro CAS:  
undefined
L/C aniline mix A (3 components)
Catalog # REST31210
Fournisseur:  Restek
Numéro CAS:  

Spécifications

  • Concentration:
    2000 μg/ml each in methylene chloride
  • Pk:
    1
  • Description:
    L/C aniline mix A
  • Pk Info:
    1 ml/ampoule

Spécifications

À propos de cet article

L/C aniline mix A contains 2-nitroaniline, 3-nitroaniline, 4-nitroaniline.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.