Cis-1,2-Dichloroethene
REST30279
: Restek
:
- Concentration:2000 μg/ml in P and T methanol
- Pk:1 mL
- CAS No.:156-59-2
- Description:cis-1,2-Dichloroethene
- Formula:C₂H₂Cl₂
- Pk Info:1 ml/ampoule
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.