Herbicide mix #4 (8 components), free acid form
Catalog # REST32061
Supplier: Restek
CAS Number:
Specifications
- Concentration:200 µg/ml each in hexane
- Pk:1 mL
- Description:Herbicide mix #4
- Pk Info:1 ml/ampoule
Specifications
About this item
This mix contains eight components.
Components are acifluorfen, bentazon, chloramben, DCPA diacid (tetrachloroterephthalic acid), 3,5-dichlorobenzoic acid, 4-nitrophenol, pentachlorophenol, picloram.
Contains: Acifluorfen (50594-66-6); Bentazon (25057-89-0); Chloramben (133-90-4); DCPA diacid (tetrachloroterephthalic acid) (2136-79-0); 3,5-Dichlorobenzoic acid (51-36-5); 4-Nitrophenol (100-02-7); Pentachlorophenol (87-86-5); Picloram (1918-02-1)
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.