Order Entry
Belgium
ContactUsLinkComponent
Herbicide mix #3 (2 components), derivatised form
Herbicide mix #3 (2 components), derivatised form
Catalog # REST32059
Supplier:  Restek
CAS Number:  
undefined
Herbicide mix #3 (2 components), derivatised form
Catalog # REST32059
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    20000 µg/ml each in hexane
  • Pk:
    1 mL
  • Description:
    Herbicide mix #3
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

This mix contains two components.

Components are MCPA methyl ester, MCPP (mecoprop) methyl ester.

Contains: MCPA methyl ester (2436-73-9); MCPP (Mecoprop) methyl ester (2786-19-8)

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.