Order Entry
Belgium
ContactUsLinkComponent
624 Surrogate standard mix (3 components)
624 Surrogate standard mix (3 components)
Catalog # REST30243
Supplier:  Restek
CAS Number:  
undefined
624 Surrogate standard mix (3 components)
Catalog # REST30243
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 μg/ml each in P and T methanol
  • Pk:
    1 mL
  • Description:
    Surrogate standard mix

Specifications

About this item

Mix contains: 1-Bromo-4-fluorobenzene (BFB) (460-00-4); Fluorobenzene (462-06-6); Pentafluorobenzene (363-72-4).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.