Herbicide mix #1 (7 components), derivatised form
Catalog # REST32055
Supplier: Restek
CAS Number:
Specifications
- Concentration:200 µg/ml each in hexane
- Pk:1 mL
- Description:Herbicide mix #1
- Pk Info:1 ml/ampoule
Specifications
About this item
This mix contains seven components.
Components are 2,4-D methyl ester, 2,4-DB methyl ester, 2,4,5-T methyl ester, 2,4,5-TP methyl ester, dicamba methyl ester, dichlorprop methyl ester, dinoseb methyl ether.
Contains: 2,4-D methyl ester (1928-38-7); 2,4-DB methyl ester (18625-12-2); 2,4,5-T methyl ester (1928-37-6); 2,4,5-TP methyl ester (Silvex) (4841-20-7); Dicamba methyl ester (6597-78-0); Dichlorprop methyl ester (57153-17-0); Dinoseb methyl ether (6099-79-2)
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.