Herbicide mix #1 (7 components), free acid form
Catalog # REST32054
Supplier: Restek
CAS Number:
Specifications
- Concentration:200 µg/ml each in hexane
- Pk:1 mL
- Description:Herbicide mix #1
- Pk Info:1 ml/ampoule
Specifications
About this item
This mix contains seven components.
Components are 2,4-D, 2,4-DB, 2,4,5-T, 2,4,5-TP, dicamba, dichlorprop, dinoseb.
Contains: 2,4-D (94-75-7); 2,4-DB (94-82-6); 2,4,5-T (93-76-5); 2,4,5-TP (Silvex) (93-72-1); Dicamba (1918-00-9); Dichlorprop (120-36-5); Dinoseb (88-85-7)
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.