8240 Alcohols mix (5 components)
Catalog # REST30214
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 µg/ml each in P and T methanol
- Pk:1 mL
- Description:Alcohols Mix, 8240
- Environmentally Preferable:
- Pk Info:1 ml/ampoule
Specifications
About this item
Mix contains: Allyl alcohol (2-propen-1-ol) (107-18-6); 2-Chloroethanol (107-07-3); Ethanol (64-17-5); Isobutyl alcohol (78-83-1); Propargyl alcohol (107-19-7).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.