Order Entry
Belgium
ContactUsLinkComponent
VOA purgeable halocarbon mix #1 (23 components)
VOA purgeable halocarbon mix #1 (23 components)
Catalog # REST30212
Supplier:  Restek
CAS Number:  
undefined
VOA purgeable halocarbon mix #1 (23 components)
Catalog # REST30212
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in P and T methanol
  • Pk:
    1 mL
  • Description:
    VOA purgeable halocarbon mix #1
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

Mix contains: Bromodichloromethane (75-27-4); Bromoform (75-25-2); Carbon tetrachloride (56-23-5); Chlorobenzene (108-90-7); 2-Chloroethyl vinyl ether (110-75-8); Chloroform (67-66-3); Dibromochloromethane (124-48-1); 1,2-Dichlorobenzene (95-50-1); 1,3-Dichlorobenzene (541-73-1); 1,4-Dichlorobenzene (106-46-7); 1,1-Dichloroethane (75-34-3); 1,2-Dichloroethane (107-06-2); 1,1-Dichloroethene (75-35-4); trans-1,2-Dichloroethene (156-60-5); 1,2-Dichloropropane (78-87-5); cis-1,3-Dichloropropene (10061-01-5); trans-1,3-Dichloropropene (10061-02-6); Methylene chloride (dichloromethane) (75-09-2); 1,1,2,2-Tetrachloroethane (79-34-5); Tetrachloroethene (127-18-4); 1,1,1-Trichloroethane (71-55-6); 1,1,2-Trichloroethane (79-00-5); Trichloroethene (79-01-6).

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.