Order Entry
Belgium
Orders LinkContactUsLinkComponent
3-Methylphenol

3-Methylphenol

3-Methylphenol, 1000 µg/ml in methanol, 1 ml/ampoule, 1 mL
Catalog # REST31403
Supplier:   Restek
3-Methylphenol
Catalog # REST31403
Supplier:  Restek
CAS Number:  
3-Methylphenol
3-Methylphenol

3-Methylphenol, 1000 µg/ml in methanol, 1 ml/ampoule, 1 mL

CAS Number:  
Price
42,80 €
Log in for availability
1 * 1 mL
Product Details & Documents

This 3-Methylphenol has minimum shelf life six months and maximum shelf life of 96 months.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.

Specifications

Specifications

  • Concentration:
    1000 µg/ml in methanol
  • Pk:
    1 mL
  • Description:
    3-Methylphenol
  • Pk Info:
    1 ml/ampoule
  • Cat. No.:
    REST31403
  • Formula:
    C₇H₈O 
  • CAS No.:
    108-39-4

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.