508 performance check mix (4 components)
REST32045
: Restek
:
- Concentration:In methyl tert-butyl ether
- Pk:1 mL
- Description:508 performance check mix
- Pk Info:1 ml/ampoule
This mix contains four components.
Components are δ-BHC (0,4 μg/ml), chlorothalonil (0,5 μg/ml), chlorpyrifos (0,02 μg/ml), DCPA methyl ester (Chlorthal-dimethyl) (0,5 μg/ml).
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.