Alaska UST method AK101AA
Alaska UST Method AK101AA (14 components), 2,000 µg/mL each in P&T methanol,, 1 mL
Catalog # REST30461
Supplier: Restek
Jump to:
Product Details & Documents
The alaska UST method consists of 14 components they are benzene, ethylbenzene, 1-ethyl-2-methylbenzene, 1-ethyl-3-methylbenzene, 1-ethyl-4-methylbenzene, isopropylbenzene (cumene), n-propylbenzene, toluene, 1,2,3-trimethylbenzene, 1,2,4-trimethylbenzene, 1,3,5-trimethylbenzene, m-xylene, o-xylene and p-xylene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.
Specifications
Specifications
- Concentration:2,000 µg/mL each in P&T methanol,
- Pk:1 mL
- Description:Alaska UST Method AK101AA (14 components)