Environmental standard, minnesota Ag list 1 pesticides mix B (6 components)
Catalog # REST32407
Supplier: Restek
CAS Number:
Specifications
- Concentration:200 ppm each in acetone
- Pk:1 mL
- Description:Minnesota Ag list 1 pesticides mix B,
- Pk Info:1 ml/ampoule
Specifications
About this item
Contains: Chlorpyrifos, EPTC, fonofos, phorate, terbufos, triallate.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfies ISO requirements.