612 Chlorinated hydrocarbons calibration mix (9 components)
N° de catalogue REST31035
Fournisseur: Restek
Numéro CAS:
Spécifications
- Concentration:2000 μg/ml each in isooctane
- Pk:1 mL
- Description:612 Chlorinated hydrocarbons calibration mix
- Pk Info:1 ml/ampoule
Spécifications
À propos de cet article
612 Chlorinated hydrocarbons calibration mix contains 2-chloronaphthalene, 1,2-dichlorobenzene, 1,3-dichlorobenzene, 1,4-dichlorobenzene, hexachlorobenzene, hexachlorobutadiene, hexachlorocyclopentadiene, hexachloroethane, 1,2,4-trichlorobenzene.
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.