Antifoam agent for purge-and-trap samples
Catalog # REST31822
Supplier: Restek
CAS Number:
Specifications
- Concentration:In Neat
- Pk:1 Ampoul
- Description:Antifoam agent for purge-and-trap
- Pk Info:1 ml/vial
Specifications
About this item
Foam generated as purge gas passes through a sample can enter the analytical trap and possibly the GC column. This silica-containing antifoam agent is effective over a wide pH range and will not conflict with chromatography of target analytes.
- Efficiently controls foam
- No hazardous materials, no components that are target analytes
- Effective at less than 0,1% of sample volume