Michigan GRO mix (14 components)
REST30468
: Restek
:
- Concentration:2000 µg/ml each in P and T methanol
- Pk:1 mL
- Description:Michigan GRO mix
- Pk Info:1 ml/ampoule
It consists of benzene, 1,2-dibromoethane, 1,2-dichloroethane, ethylbenzene, isopropylbenzene (cumene), 2-methylnaphthalene, methyl tert-butyl-ether (MTBE), naphthalene, toluene, 1,2,4-trimethylbenzene, 1,3,5-trimethylbenzene, m-xylene, o-xylene and p-xylene.
: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy ISO requirements.