611 Haloethers calibration mix (5 components)
Catalog # REST31034
Supplier: Restek
CAS Number:
Specifications
- Concentration:2000 μg/ml each in acetone
- Pk:1
- Description:611 Haloethers calibration mix
- Pk Info:1 ml/ampoule
Specifications
About this item
Haloethers calibration mix contains bis(2-chloroethoxy)methane, bis(2-chloroethyl)ether, 4-bromophenyl phenyl ether, 4-chlorophenyl phenyl ether, 2,2'-oxybis(1-chloropropane).
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.