Herbicide reference standard, 2,4,5-TP (Silvex)
Catalog # REST32245
Supplier: Restek
CAS Number:
Specifications
- Concentration:1000 μg/ml in methanol
- Pk:1
- CAS No.:93-72-1
- Description:2,4,5-TP (Silvex)
- Environmentally Preferable:
- Formula:C₉H₇Cl₃O₃
- Description:2,4,5-TP (Silvex)
- Pk Info:1 ml/ampoule
Specifications
About this item
Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.