Order Entry
Belgium
ContactUsLinkComponent
607 Nitrosamines calibration mix (3 components)
607 Nitrosamines calibration mix (3 components)
Catalog # REST31032
Supplier:  Restek
CAS Number:  
undefined
607 Nitrosamines calibration mix (3 components)
Catalog # REST31032
Supplier:  Restek
CAS Number:  

Specifications

  • Concentration:
    2000 µg/ml each in methanol
  • Pk:
    1
  • Description:
    607 Nitrosamines Calibration Mix
  • Pk Info:
    1 ml/ampoule

Specifications

About this item

607 Nitrosamines calibration mix contains N-nitrosodimethylamine, N-nitroso-di-n-propylamine, N-nitrosodiphenylamine.

Certifications: Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.