Order Entry
Belgium
ContactUsLinkComponent
Pesticide reference standard, Propachlor
Pesticide reference standard, Propachlor
  REST32235
 :  Restek
 :  
undefined
Pesticide reference standard, Propachlor
  REST32235
 :  Restek
 :  

 

  • Concentration:
    1000 μg/ml in methanol
  • Pack type:
    Glass bottle
  • Pk:
    1
  • CAS No.:
    1918-16-7
  • Description:
    Propachlor
  • Formula:
    C₁₁H₁₄ClNO
  • Description:
    Propachlor

 

 

 : Certified reference materials (CRMs) manufactured and QC-tested in ISO-accredited labs satisfy your ISO requirements.