Residual range calibration verification standard (CVS)
Catalog # REST31818
Supplier: Restek
CAS Number:
Specifications
- Concentration:25000 µg/ml in methylene chloride
- Pk:1 Ampoul
- Description:Residual range calibration verification standard (CVS)
- Pk Info:1 ml
Specifications
About this item
SAE30 motor oil:SAE40 motor oil (1:1).
- 2 components